5-(2-Furyl)-1H-pyrazole-3-carboxylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(Furan-2-yl)-1H-pyrazole-3-carboxylic acid

B25737.03 B25737.06
ALFAB25737.03EA 0 SGD
ALFAB25737.03 ALFAB25737.06
5-(2-Furyl)-1H-pyrazole-3-carboxylic acid 97%
5-(2-Furyl)-1H-pyrazole-3-carboxylic acid

Purity: 97% CAS No.: 116153-81-2 Molecular Formula: C8H6N2O3 Formula Weight: 178.15 Melting Point: 219-221° MDL No.: MFCD05170057

Formula: C₈H₆N₂O₃
MW: 178,15 g/mol
Melting Pt: 219…221 °C
Storage Temperature: Ambient
MDL Number: MFCD05170057
CAS Number: 116153-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR