1-Benzhydryl-3-azetidinol hydrochloride 95%

Supplier: Thermo Scientific
Warning

Synonyms: 1-(Diphenylmethyl)-3-hydroxyazetidine hydrochloride, 1-Benzhydrylazetidin-3-ol hydrochloride

H25819.06 H25819.03
ALFAH25819.06EA 0 SGD
ALFAH25819.06 ALFAH25819.03
1-Benzhydryl-3-azetidinol hydrochloride 95%
1-Benzhydryl-3-azetidinol hydrochloride

Purity: 95% CAS No.: 90604-02-7 Molecular Formula: C16H17NO.HCl Formula Weight: 275.78 Melting Point: 172-174° MDL No.: MFCD00209447

Formula: C₁₆H₁₇NO·HCl
MW: 275,78 g/mol
Melting Pt: 172…174 °C
Storage Temperature: Ambient
MDL Number: MFCD00209447
CAS Number: 90604-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR