Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine 94%

Supplier: Thermo Scientific
Danger

H25882.06 H25882.03
ALFAH25882.06EA 0 SGD
ALFAH25882.06 ALFAH25882.03
Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine 94%
Bis(3,5-di-tert-butyl-4-methoxyphenyl)chlorophosphine

Purity: 94% CAS No.: 212713-08-1 Molecular Formula: C30H46ClO2P Formula Weight: 505.13 UN No.: UN3261 MDL No.: MFCD08458881

Formula: C₃₀H₄₆ClO₂P
MW: 505,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08458881
CAS Number: 212713-08-1
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR