6,7-Dihydro-5H-cyclopenta[b]pyridine 99%

Supplier: Thermo Scientific
Warning

Synonyms: 5H-1-Pyrindine, 6,7-dihydro-, 2,3-Cyclopentenopyridine

H25896.06 H25896.14
ALFAH25896.06EA 125 SGD
ALFAH25896.06 ALFAH25896.14
6,7-Dihydro-5H-cyclopenta[b]pyridine 99%
6,7-Dihydro-5H-cyclopenta[b]pyridine

Purity: 99% CAS No.: 533-37-9 Molecular Formula: C8H9N Formula Weight: 119.17 Flash Point: 67°(153°F) Refractive Index: 1.5440 Boiling Point: 87-88°/11mm MDL No.: MFCD00005933

Formula: C₈H₉N
MW: 119,17 g/mol
Boiling Pt: 87…88 °C (11 mmHg)
Density: 1,036
Flash Pt: 67 °C (153 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005933
CAS Number: 533-37-9
EINECS: 208-564-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR