(2-Chloro-4-methoxyphenyl)acetonitrile 95%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
2-(2-Chloro-4-methoxyphenyl)acetonitrile
H26021.03
H26021.06
ALFAH26021.03EA
0
SGD
ALFAH26021.03
ALFAH26021.06
(2-Chloro-4-methoxyphenyl)acetonitrile 95%
(2-Chloro-4-methoxyphenyl)acetonitrile
Purity: 95%
CAS No.: 170737-93-6
Molecular Formula: C9H8ClNO
Formula Weight: 181.62
Melting Point: 41-43°
UN No.: UN3439
MDL No.: MFCD08445710
Formula:
C₉H₈ClNO MW: 181,62 g/mol Melting Pt: 41…43 °C Storage Temperature: Ambient |
MDL Number:
MFCD08445710 CAS Number: 170737-93-6 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...