2,3-Diamino-6-methoxypyridine dihydrochloride 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-Methoxypyridine-2,3-diamine dihydrochloride
H26472.MD
H26472.03
ALFAH26472.MDEA
0
SGD
ALFAH26472.MD
ALFAH26472.03
2,3-Diamino-6-methoxypyridine dihydrochloride 96%
2,3-Diamino-6-methoxypyridine dihydrochloride
Purity: 96%
CAS No.: 94166-62-8
Molecular Formula: C6H9N3O.2HCl
Formula Weight: 212.08
Melting Point: 167-171°
MDL No.: MFCD07787253
Formula:
C₆H₁₁Cl₂N₃O MW: 212,074 g/mol Melting Pt: 167…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00137799 CAS Number: 94166-62-8 EINECS: None |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...