3-Benzylamino-1-propanol 97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 3-(Benzylamino)propan-1-ol, N-(3-Hydroxypropyl)benzylamine, 3-(Benzylamino)-1-propanol

H26664.MD H26664.03
ALFAH26664.MDEA 0 SGD
ALFAH26664.MD ALFAH26664.03
3-Benzylamino-1-propanol 97%
3-Benzylamino-1-propanol

Purity: 97% CAS No.: 4720-29-0 Molecular Formula: C6H5CH2NH(CH2)3OH Formula Weight: 165.24 MDL No.: MFCD00020622

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00020622
CAS Number: 4720-29-0
EINECS: 225-210-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR