1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

Supplier: Thermo Scientific
Warning

H26674.MD H26674.03
ALFAH26674.MDEA 182 SGD
ALFAH26674.MD ALFAH26674.03
1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane
1,4,8,11-Tetraazatricyclo[9.3.1.1(4,8)]hexadecane

CAS No.: 75920-10-4 Molecular Formula: C12H24N4 Formula Weight: 224.35 Melting Point: 83-85° MDL No.: MFCD09953472

Formula: C₁₂H₂₄N₄
MW: 224,35 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD09953472
CAS Number: 75920-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR