2,2'-Dibromo-1,1'-binaphthyl 96%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
racemic-2,2'-Dibromo-1,1'-binaphthyl, 2,2-Dibromo-1,1-binaphthyl, 2,2'-Dibromo-1,1'-binaphthalene
H26987.03
H26987.06
ALFAH26987.03EA
0
SGD
ALFAH26987.03
ALFAH26987.06
2,2'-Dibromo-1,1'-binaphthyl 96%
2,2'-Dibromo-1,1'-binaphthyl
Purity: 96%
CAS No.: 74866-28-7
Molecular Formula: C20H12Br2
Formula Weight: 412.23
Boiling Point: 178-183°
MDL No.: MFCD00188005
Formula:
C₂₀H₁₂Br₂ MW: 412,12 g/mol Boiling Pt: 178…183 °C Storage Temperature: Ambient |
MDL Number:
MFCD00188005 CAS Number: 74866-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...