(+/-)-1-Benzyloxycarbonyl-4-boc-piperazine-2-carboxylic acid 97%

Supplier: Thermo Scientific
Warning

H27047.MD H27047.03 H27047.06
ALFAH27047.MDEA 0 SGD
ALFAH27047.MD ALFAH27047.03 ALFAH27047.06
(+/-)-1-Benzyloxycarbonyl-4-boc-piperazine-2-carboxylic acid 97%
(+/-)-1-Benzyloxycarbonyl-4-boc-piperazine-2-carboxylic acid

Purity: 97% CAS No.: 126937-41-5 Molecular Formula: C18H24N2O6 Formula Weight: 364.40 Melting Point: 150-152° MDL No.: MFCD01631192

Formula: C₁₈H₂₄N₂O₆
MW: 364,40 g/mol
Melting Pt: 150…152 °C
Storage Temperature: Ambient
MDL Number: MFCD01631192
CAS Number: 126937-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR