1-(1-Boc-3-azetidinyl)pyrrolidine 94%
Supplier: Thermo Scientific
|
Warning
|
H27111.MD
H27111.03
ALFAH27111.MDEA
0
SGD
ALFAH27111.MD
ALFAH27111.03
1-(1-Boc-3-azetidinyl)pyrrolidine 94%
1-(1-Boc-3-azetidinyl)pyrrolidine
Purity: 94%
CAS No.: 1019008-21-9
Molecular Formula: C12H22N2O2
Formula Weight: 226.32
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4780
MDL No.: MFCD09860616
Formula:
C₁₂H₂₂N₂O₂ MW: 226,32 g/mol |
MDL Number:
MFCD09860616 CAS Number: 1019008-21-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...