2-Chloroquinoline-3-carbaldehyde 98%

Supplier: Thermo Scientific
Warning

H27133.09 H27133.03
ALFAH27133.09EA 353 SGD
ALFAH27133.09 ALFAH27133.03
2-Chloroquinoline-3-carbaldehyde 98%
2-Chloroquinoline-3-carbaldehyde

Purity: 98% CAS No.: 73568-25-9 Molecular Formula: C10H6ClNO Formula Weight: 191.62 Melting Point: 148-150° MDL No.: MFCD00130079

Formula: C₁₀H₆ClNO
MW: 191,62 g/mol
Melting Pt: 148…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00130079
CAS Number: 73568-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR