2-(Methylsulphinyl)benzenecarboxylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(Methylsulphinyl)benzoic acid

H27144.03 H27144.06
ALFAH27144.03EA 0 SGD
ALFAH27144.03 ALFAH27144.06
2-(Methylsulphinyl)benzenecarboxylic acid 97%
2-(Methylsulphinyl)benzenecarboxylic acid

Purity: 97% CAS No.: 19093-34-6 Molecular Formula: C8H8O3S Formula Weight: 184.21 Melting Point: 150-160° MDL No.: MFCD01936003

Formula: C₈H₈O₃S
MW: 184,22 g/mol
Melting Pt: 150…160 °C
Storage Temperature: Ambient
MDL Number: MFCD01936003
CAS Number: 19093-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR