3,5-Dibromo-4-cyanopyridine 97%

Supplier: Thermo Scientific
Danger

Synonyms: 3,5-Dibromoisonicotinonitrile

H27186.MD H27186.03
ALFAH27186.MDEA 0 SGD
ALFAH27186.MD ALFAH27186.03
3,5-Dibromo-4-cyanopyridine 97%
3,5-Dibromo-4-cyanopyridine

Purity: 97% CAS No.: 870244-34-1 Molecular Formula: C6H2Br2N2 Formula Weight: 261.90 Melting Point: 149-151° UN No.: UN3439 MDL No.: MFCD09263321

Formula: C₆H₂Br₂N₂
MW: 261,90 g/mol
Melting Pt: 149…151 °C
Storage Temperature: Ambient
MDL Number: MFCD09263321
CAS Number: 870244-34-1
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR