5-(Trimethylsilyl)-1,3-cyclopentadiene mixture of isomers 90%, tech.
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Trimethylsilylcyclopentadiene (mixture of isomers), (2,4-Cyclopentadien-1-yl)trimethylsilane
H27192.03
H27192.09
ALFAH27192.03EA
0
SGD
ALFAH27192.03
ALFAH27192.09
5-(Trimethylsilyl)-1,3-cyclopentadiene mixture of isomers 90%, tech.
5-(Trimethylsilyl)-1,3-cyclopentadiene
Purity: 90%
CAS No.: 3559-74-8
Molecular Formula: C8H14Si
Formula Weight: 138.28
Storage Temperature: -30°C to -10°C
Flash Point: 29°(84°F)
Boiling Point: 138-140°
UN No.: UN1993
MDL No.: MFCD00010666
Formula:
C₈H₁₄Si MW: 138,29 g/mol Boiling Pt: 138…140 °C Density: 0,833 Flash Pt: 29 °C (84 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00010666 CAS Number: 3559-74-8 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...