4-Cyano-3-iodopyridine 98%

Supplier: Thermo Scientific
Warning

H27247.MD H27247.03
ALFAH27247.MDEA 0 SGD
ALFAH27247.MD ALFAH27247.03
4-Cyano-3-iodopyridine 98%
4-Cyano-3-iodopyridine

Purity: 98% CAS No.: 10386-28-4 Molecular Formula: C6H3IN2 Formula Weight: 230.01 Melting Point: 119-122° UN No.: UN3439 MDL No.: MFCD08729285

Formula: C₆H₃IN₂
MW: 230,01 g/mol
Melting Pt: 119…122 °C
Storage Temperature: Ambient
MDL Number: MFCD08729285
CAS Number: 10386-28-4
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR