1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%

Supplier: Thermo Fisher Scientific

Synonyms: 1-Butyl-4-((4-methoxyphenyl)ethynyl)benzene

H27304.09 H27304.03
ALFAH27304.09EA 0 SGD
ALFAH27304.09 ALFAH27304.03
1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene ≥99%
1-n-Butyl-4-[(4-methoxyphenyl)ethynyl]benzene

Purity: 99+% CAS No.: 35684-12-9 Molecular Formula: C19H20O Formula Weight: 264.37 Melting Point: 40°(N) Boiling Point: 172-172°/1mm MDL No.: MFCD04038796

Formula: C₁₉H₂₀O
MW: 264.37 g/mol
Boiling Pt: 172…172 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD04038796
CAS Number: 35684-12-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR