(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
Supplier: Thermo Scientific
|
Warning
|
H27321.MD
H27321.03
ALFAH27321.MDEA
0
SGD
ALFAH27321.MD
ALFAH27321.03
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine
CAS No.: 61478-28-2
Molecular Formula: C34H37NO2P2
Formula Weight: 553.62
Storage Temperature: 2°C to 8°C
Melting Point: 100-103°
MDL No.: MFCD01631275
Formula:
C₃₄H₃₇NO₂P₂ MW: 553,62 g/mol Melting Pt: 100…103 °C |
MDL Number:
MFCD01631275 CAS Number: 61478-28-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...