4,6-Bis(diphenylphosphino)phenoxazine ≥98%

Supplier: Thermo Scientific

Synonyms: 4,6-Bis(diphenylphosphino)-10H-phenoxazine, NIXANTPHOS

H27330.MD H27330.03 H27330.06
ALFAH27330.MDEA 200 SGD
ALFAH27330.MD ALFAH27330.03 ALFAH27330.06
4,6-Bis(diphenylphosphino)phenoxazine ≥98%
4,6-Bis(diphenylphosphino)phenoxazine

Purity: 98+% CAS No.: 261733-18-0 Molecular Formula: C36H27NOP2 Formula Weight: 551.55 Melting Point: 251-254° MDL No.: MFCD03788937

Formula: C₃₆H₂₇NOP₂
MW: 551,57 g/mol
Melting Pt: 251…254 °C
Storage Temperature: Ambient
MDL Number: MFCD03788937
CAS Number: 261733-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR