6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 95%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(Pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(Pyrrolidino)pyridine-5-boronic acid pinacol ester

H27341.MD H27341.03
ALFAH27341.MDEA 0 SGD
ALFAH27341.MD ALFAH27341.03
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester 95%
6-(1-Pyrrolidinyl)pyridine-3-boronic acid pinacol ester

Purity: 95% CAS No.: 933986-97-1 Molecular Formula: C15H23BN2O2 Formula Weight: 274.17 Melting Point: 118-121° MDL No.: MFCD06798273

Formula: C₁₅H₂₃BN₂O₂
MW: 274,17 g/mol
Melting Pt: 118…121 °C
Storage Temperature: Ambient
MDL Number: MFCD06798273
CAS Number: 933986-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR