4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene 99%
Supplier: Thermo Scientific
Synonyms:
9,9-Dimethyl-4,5-bis(di-t-butylphosphino)xanthene, t-Bu-Xantphos, (9,9-Dimethyl-9H-xanthene-4,5-diyl)bis(di-tert-butylphosphine)
H27420.ME
H27420.03
H27420.06
ALFAH27420.MEEA
0
SGD
ALFAH27420.ME
ALFAH27420.03
ALFAH27420.06
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene 99%
4,5-Bis(di-tert-butylphosphino)-9,9-dimethylxanthene
Purity: 99%
Molecular Formula: C31H48OP2
Formula Weight: 498.68
Melting Point: 153-157°
MDL No.: MFCD07781995
Formula:
C₃₁H₄₈OP₂ MW: 498,68 g/mol Melting Pt: 153…157 °C Storage Temperature: Ambient |
MDL Number:
MFCD07781995 CAS Number: 856405-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...