Methyl pyridazine-4-carboxylate 96%
Supplier: Thermo Scientific
H27449.03
H27449.06
ALFAH27449.03EA
0
SGD
ALFAH27449.03
ALFAH27449.06
Methyl pyridazine-4-carboxylate 96%
Methyl pyridazine-4-carboxylate
Purity: 96%
CAS No.: 34231-77-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Melting Point: 64-67°
MDL No.: MFCD09953488
Formula:
C₆H₆N₂O₂ MW: 138,13 g/mol Melting Pt: 64…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD09953488 CAS Number: 34231-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...