N-Methylbutan-2-amine hydrochloride 94%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(Methylamino)butane hydrochloride, 2-Methylamino-butane-hydrochloride

H27547.06 H27547.03
ALFAH27547.06EA 498 SGD
ALFAH27547.06 ALFAH27547.03
N-Methylbutan-2-amine hydrochloride 94%
N-Methylbutan-2-amine hydrochloride

Purity: 94% CAS No.: 75098-40-7 Molecular Formula: C5H13N.HCl Formula Weight: 123.62 Melting Point: 76-82° MDL No.: MFCD07777098

Formula: C₅H₁₃N·HCl
MW: 123,62 g/mol
Melting Pt: 76…82 °C
Storage Temperature: Ambient
MDL Number: MFCD07777098
CAS Number: 75098-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR