4-(Dibenzylamino)benzeneboronic acid ≥95%

Supplier: Thermo Scientific
Warning

H27550.MD H27550.03
ALFAH27550.MDEA 0 SGD
ALFAH27550.MD ALFAH27550.03
4-(Dibenzylamino)benzeneboronic acid ≥95%
4-(Dibenzylamino)benzeneboronic acid

Purity: 95% CAS No.: 159191-44-3 Molecular Formula: C20H20BNO2 Formula Weight: 317.19 Melting Point: 184-188° MDL No.: MFCD06798225

Formula: C₂₀H₂₀BNO₂
MW: 317,19 g/mol
Melting Pt: 184…188 °C
Storage Temperature: Ambient
MDL Number: MFCD06798225
CAS Number: 159191-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR