Methyl-5-formylthiophene-3-carboxylate 95%

Supplier: Thermo Scientific
Warning

H27597.03 H27597.06
ALFAH27597.03EA 364 SGD
ALFAH27597.03 ALFAH27597.06
Methyl-5-formylthiophene-3-carboxylate 95%
Methyl-5-formylthiophene-3-carboxylate

Purity: 95% CAS No.: 67808-66-6 Molecular Formula: C7H6O3S Formula Weight: 170.19 Melting Point: 113-117° MDL No.: MFCD01859945

Formula: C₇H₆O₃S
MW: 170,19 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD01859945
CAS Number: 67808-66-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR