(R)-1-(2-Methoxyphenyl)ethylamine ≥99%, ee 98%, ChiPros®

Supplier: Thermo Scientific
Danger

Synonyms: (R)-1-(2-Methoxyphenyl)ethanamine

H27669.03 H27669.06
ALFAH27669.03EA 0 SGD
ALFAH27669.03 ALFAH27669.06
(R)-1-(2-Methoxyphenyl)ethylamine ≥99%, ee 98%, ChiPros®
(R)-1-(2-Methoxyphenyl)ethylamine

Purity: 99% CAS No.: 68285-23-4 Molecular Formula: C9H13NO Formula Weight: 151.21 Boiling Point: 118°/17mm UN No.: UN2735 MDL No.: MFCD00671657

Formula: C₉H₁₃NO
MW: 151,21 g/mol
Boiling Pt: 118 °C (17 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00671657
CAS Number: 68285-23-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR