Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Bis(t-butylcyclopentadienyl)titanium dichloride

H27714.03 H27714.06
ALFAH27714.03EA 0 SGD
ALFAH27714.03 ALFAH27714.06
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride ≥98%
Bis(tert-butylcyclopentadienyl)titanium(IV) dichloride

Purity: 98+% CAS No.: 79269-71-9 Molecular Formula: C18H26Cl2Ti Formula Weight: 361.17 Melting Point: 233° MDL No.: MFCD01073774

Formula: C₁₈H₂₆Cl₂Ti
MW: 361,172 g/mol
Melting Pt: 233 °C
Storage Temperature: Ambient
MDL Number: MFCD01073774
CAS Number: 79269-71-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR