N-Boc-4-methylene-L-proline 97%
Supplier: Thermo Scientific
Synonyms:
(S)-1-(tert-Butoxycarbonyl)-4-methylenepyrrolidine-2-carboxylic acid
H27746.MD
H27746.03
H27746.06
ALFAH27746.MDEA
0
SGD
ALFAH27746.MD
ALFAH27746.03
ALFAH27746.06
N-Boc-4-methylene-L-proline 97%
N-Boc-4-methylene-L-proline
Purity: 97%
CAS No.: 84348-38-9
Molecular Formula: C11H17NO4
Formula Weight: 227.26
Storage Temperature: 2°C to 8°C
Melting Point: 110-114°
MDL No.: MFCD01861787
Formula:
C₁₁H₁₇NO₄ MW: 227,26 g/mol Melting Pt: 110…114 °C |
MDL Number:
MFCD01861787 CAS Number: 84348-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...