1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%

Supplier: Thermo Scientific
Danger

H27762.06 H27762.03
ALFAH27762.06EA 0 SGD
ALFAH27762.06 ALFAH27762.03
1-Fluoro-3,5-dimethyl-2-nitrobenzene 98%
1-Fluoro-3,5-dimethyl-2-nitrobenzene

Purity: 98% CAS No.: 315-13-9 Molecular Formula: C8H8FNO2 Formula Weight: 169.16 Melting Point: 54-57° MDL No.: MFCD00052367

Formula: C₈H₈FNO₂
MW: 169,16 g/mol
Melting Pt: 54…57 °C
Storage Temperature: Ambient
MDL Number: MFCD00052367
CAS Number: 315-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR