5-Bromo-2-phenylpyridine 95%

Supplier: Thermo Scientific
Warning

H27816.03 H27816.MD
ALFAH27816.03EA 0 SGD
ALFAH27816.03 ALFAH27816.MD
5-Bromo-2-phenylpyridine 95%
5-Bromo-2-phenylpyridine

Purity: 95% CAS No.: 27012-25-5 Molecular Formula: C11H8BrN Formula Weight: 234.10 Melting Point: 73-75° MDL No.: MFCD04114181

Formula: C₁₁H₈BrN
MW: 234,10 g/mol
Melting Pt: 73…75 °C
Storage Temperature: Ambient
MDL Number: MFCD04114181
CAS Number: 27012-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR