(1S,2S)-N-Methylsulphonyl-1,2-diphenylethanediamine ≥98%
Supplier: Thermo Scientific
|
Danger
|
H27867.03
H27867.06
ALFAH27867.03EA
362
SGD
ALFAH27867.03
ALFAH27867.06
(1S,2S)-N-Methylsulphonyl-1,2-diphenylethanediamine ≥98%
(1S,2S)-N-Methylsulphonyl-1,2-diphenylethanediamine
Purity: 98+%
CAS No.: 300345-76-0
Molecular Formula: C15H18N2O2S
Formula Weight: 290.38
Melting Point: 117-120°
Optical Rotation: 1.8° (c=1 in chloroform)
UN No.: UN3259
MDL No.: MFCD03791037
Formula:
C₁₅H₁₈N₂O₂S Melting Pt: 117…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791037 CAS Number: 300345-76-0 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...