Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Supplier: Thermo Scientific
Warning
|
Synonyms:
Pentamethylcyclopentadienyltitanium trimethoxide
H27885.03
H27885.06
ALFAH27885.03EA
0
SGD
ALFAH27885.03
ALFAH27885.06
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV) ≥97%
Trimethoxy(pentamethylcyclopentadienyl)titanium(IV)
Purity: 97+%
CAS No.: 123927-75-3
Molecular Formula: C13H24O3Ti
Formula Weight: 276.21
Flash Point: 61°(142°F)
Refractive Index: 1.554
Boiling Point: 285°
MDL No.: MFCD00269850
Formula:
C₁₃H₂₄O₃Ti MW: 276,199 g/mol Boiling Pt: 285 °C Density: 1,081 Flash Pt: 61 °C (142 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00269850 CAS Number: 123927-75-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...