(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97%

Supplier: Thermo Scientific

Synonyms: 1,2-Bis((S)-(2-methoxyphenyl)(phenyl)phosphino)ethane, (S,S)-Ethylenebis[(2-methoxyphenyl)phenylphosphine], (S,S)-(+)-1,2-Bis[(2-methoxyphenyl)(phenyl)phosphino]ethane, (+)-DIPAMP, (S,S)-DIPAMP

H28037.MD H28037.03 H28037.MC
ALFAH28037.MDEA 0 SGD
ALFAH28037.MD ALFAH28037.03 ALFAH28037.MC
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane 97%
(S,S)-1,2-Bis[(2-methoxyphenyl)phenylphosphino]ethane

Purity: 97% CAS No.: 97858-62-3 Molecular Formula: C28H28O2P2 Formula Weight: 458.47 Melting Point: 101-105° Optical Rotation: +84° (c=1 in chloroform) MDL No.: MFCD05863547

Formula: C₂₈H₂₈O₂P₂
MW: 458,48 g/mol
Melting Pt: 101…105 °C
Storage Temperature: Ambient
MDL Number: MFCD05863547
CAS Number: 97858-62-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR