4-Amino-1H-pyrazole 95%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Aminopyrazole, 1H-Pyrazol-4-amine

H28921.ME H28921.MC
ALFAH28921.MEEA 325 SGD
ALFAH28921.ME ALFAH28921.MC
4-Amino-1H-pyrazole 95%
4-Amino-1H-pyrazole

Purity: 95% CAS No.: 28466-26-4 Molecular Formula: C3H5N3 Formula Weight: 83.09 Melting Point: 72-77° MDL No.: MFCD01693729

Formula: C₃H₅N₃
MW: 83,09 g/mol
Melting Pt: 72…77 °C
Storage Temperature: Ambient
MDL Number: MFCD01693729
CAS Number: 28466-26-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR