2,2-Difluorocyclopropylamine hydrochloride 97%

Supplier: Thermo Scientific
Warning

H29051.ME H29051.MC
ALFAH29051.MEEA 0 SGD
ALFAH29051.ME ALFAH29051.MC
2,2-Difluorocyclopropylamine hydrochloride 97%
2,2-Difluorocyclopropylamine hydrochloride

Purity: 97% CAS No.: 105614-25-3 Molecular Formula: C3H5F2N.HCl Formula Weight: 129.54 Melting Point: ca 145° dec. MDL No.: MFCD07777150

Formula: C₃H₆ClF₂N
MW: 129,54 g/mol
Melting Pt: ∼ 145 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD07777150
CAS Number: 105614-25-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR