4-Bromo-2-nitrophenylhydrazine hydrochloride 95%
Supplier: Thermo Scientific
|
Danger
|
H29202.03
H29202.06
ALFAH29202.03EA
0
SGD
ALFAH29202.03
ALFAH29202.06
4-Bromo-2-nitrophenylhydrazine hydrochloride 95%
4-Bromo-2-nitrophenylhydrazine hydrochloride
Purity: 95%
CAS No.: 100032-79-9
Molecular Formula: C6H6BrN3O2.HCl
Formula Weight: 268.50
Melting Point: ca 185° dec.
MDL No.: MFCD09260482
Formula:
C₆H₇BrClN₃O₂ Melting Pt: ∼ 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD09260482 CAS Number: 100032-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...