Dimethyl oxazole-4,5-dicarboxylate 99%

Supplier: Thermo Scientific

H30009.03 H30009.09
ALFAH30009.03EA 0 SGD
ALFAH30009.03 ALFAH30009.09
Dimethyl oxazole-4,5-dicarboxylate 99%
Dimethyl oxazole-4,5-dicarboxylate

Purity: 99% CAS No.: 72030-81-0 Molecular Formula: C7H7NO5 Formula Weight: 185.14 Melting Point: 74-76° Boiling Point: 150°/12mm MDL No.: MFCD05863258

Formula: C₇H₇NO₅
MW: 185,14 g/mol
Boiling Pt: 150 °C (12 mmHg)
Melting Pt: 74…76 °C
Storage Temperature: Ambient
MDL Number: MFCD05863258
CAS Number: 72030-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR