4-Ethyl-trans-β-styrylboronic acid pinacol ester 97%

Supplier: Thermo Scientific

H30744.MD H30744.03
ALFAH30744.MDEA 0 SGD
ALFAH30744.MD ALFAH30744.03
4-Ethyl-trans-β-styrylboronic acid pinacol ester 97%
4-Ethyl-trans-beta-styrylboronic acid pinacol ester

Purity: 97% CAS No.: 870717-91-2 Molecular Formula: C16H23BO2 Formula Weight: 258.17 Flash Point: 110°(230°F) Melting Point: 46-48° MDL No.: MFCD08276837

Formula: C₁₆H₂₃BO₂
MW: 258,17 g/mol
Melting Pt: 46…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD08276837
CAS Number: 870717-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR