(2-Allylphenoxy)trimethylsilane ≥98%

Supplier: Thermo Scientific
Warning

H30905.06 H30905.14 H30905.22
ALFAH30905.06EA 0 SGD
ALFAH30905.06 ALFAH30905.14 ALFAH30905.22
(2-Allylphenoxy)trimethylsilane ≥98%
(2-Allylphenoxy)trimethylsilane

Purity: 98% CAS No.: 18042-43-8 Molecular Formula: C12H18OSi Formula Weight: 206.36 Boiling Point: 85°/5mm MDL No.: MFCD12407078

Formula: C₁₂H₁₈OSi
MW: 206,36 g/mol
Boiling Pt: 85 °C (5 mmHg)
Density: 0,925
Storage Temperature: Ambient
MDL Number: MFCD12407078
CAS Number: 18042-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR