1-(4-Methoxyphenyl)-1H-1,2,4-triazole 99%

Supplier: Thermo Scientific
Warning

H30962.06 H30962.14
ALFAH30962.06EA 0 SGD
ALFAH30962.06 ALFAH30962.14
1-(4-Methoxyphenyl)-1H-1,2,4-triazole 99%
1-(4-Methoxyphenyl)-1H-1,2,4-triazole

Purity: 99% CAS No.: 68377-33-3 Molecular Formula: C9H9N3O Formula Weight: 175.19 Melting Point: 96-98° MDL No.: MFCD12407079

Formula: C₉H₉N₃O
MW: 175,19 g/mol
Melting Pt: 96…98 °C
Storage Temperature: Ambient
MDL Number: MFCD12407079
CAS Number: 68377-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR