Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester 97%
Supplier: Thermo Scientific
|
Warning
|
H31355.MD
H31355.03
ALFAH31355.MDEA
0
SGD
ALFAH31355.MD
ALFAH31355.03
Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester 97%
Tetrahydrothiophene-1,1-dioxide-3-boronic acid diethanolamine ester
Purity: 97%
Molecular Formula: C8H16BNO4S
Formula Weight: 233.10
Melting Point: 242-244°
MDL No.: MFCD12546196
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...