7-Amino-2H-1,4-benzoxazin-3(4H)-one 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
7-Amino-2H-1,4-benzoxazin-3[4H]one
H31534.MD
H31534.03
ALFAH31534.MDEA
119
SGD
ALFAH31534.MD
ALFAH31534.03
7-Amino-2H-1,4-benzoxazin-3(4H)-one 97%
7-Amino-2H-1,4-benzoxazin-3(4H)-one
Purity: 97%
CAS No.: 26215-14-5
Molecular Formula: C8H8N2O2
Formula Weight: 164.17
Physical Form: Crystalline needles
Appearance: Pale yellow to orange
Melting Point: 220-222°
MDL No.: MFCD03425794
Formula:
C₈H₈N₂O₂ MW: 164,17 g/mol Melting Pt: 220…222 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425794 CAS Number: 26215-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...