5-(4-Fluorophenyl)-2-thiophenecarboxylic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 5-(4-Fluorophenyl)thiophene-2-carboxylic acid

H31579.03 H31579.06
ALFAH31579.03EA 0 SGD
ALFAH31579.03 ALFAH31579.06
5-(4-Fluorophenyl)-2-thiophenecarboxylic acid 97%
5-(4-Fluorophenyl)-2-thiophenecarboxylic acid

Purity: 97% CAS No.: 115933-30-7 Molecular Formula: C11H7FO2S Formula Weight: 222.24 Melting Point: 250-252° MDL No.: MFCD01316570

Formula: C₁₁H₇FO₂S
MW: 222,24 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD01316570
CAS Number: 115933-30-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR