[1,2,4]Triazol-1-ylacetic acid 97%

Supplier: Thermo Scientific
Warning

Synonyms: 2-(1H-1,2,4-Triazol-1-yl)acetic acid

H31732.MD H31732.03
ALFAH31732.MDEA 181 SGD
ALFAH31732.MD ALFAH31732.03
[1,2,4]Triazol-1-ylacetic acid 97%
[1,2,4]Triazol-1-ylacetic acid

Purity: 97% CAS No.: 28711-29-7 Molecular Formula: C4H5N3O2 Formula Weight: 127.10 Melting Point: 201-205° MDL No.: MFCD00036051

Formula: C₄H₅N₃O₂
MW: 127,1 g/mol
Melting Pt: 201…205 °C
Storage Temperature: Ambient
MDL Number: MFCD00036051
CAS Number: 28711-29-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR