2-(2-Furyl)-1,3-diphenylimidazolidine 96%
Supplier: Thermo Scientific
|
Warning
|
H31757.MD
H31757.03
ALFAH31757.MDEA
0
SGD
ALFAH31757.MD
ALFAH31757.03
2-(2-Furyl)-1,3-diphenylimidazolidine 96%
2-(2-Furyl)-1,3-diphenylimidazolidine
Purity: 96%
CAS No.: 94378-00-4
Molecular Formula: C19H18N2O
Formula Weight: 290.37
Melting Point: 129-131°
MDL No.: MFCD00225139
Formula:
C₁₉H₁₈N₂O MW: 290,37 g/mol Melting Pt: 129…131 °C Storage Temperature: Ambient |
MDL Number:
MFCD00225139 CAS Number: 94378-00-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...