trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone 96%

Supplier: Thermo Scientific
Warning

H31932.MD H31932.03
ALFAH31932.MDEA 0 SGD
ALFAH31932.MD ALFAH31932.03
trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone 96%
trans-2-[trans-2-(1,3,3-Trimethyl-2-indolinylidene)ethylidene]-1-indanone

Purity: 96% CAS No.: 53704-23-7 Molecular Formula: C22H21NO Formula Weight: 315.42 Melting Point: 184-187° MDL No.: MFCD06409584

Formula: C₂₂H₂₁NO
MW: 315,42 g/mol
Melting Pt: 184…187 °C
Storage Temperature: Ambient
MDL Number: MFCD06409584
CAS Number: 53704-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR