1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Supplier: Thermo Scientific
Synonyms:
3-((4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)pyridine
H31954.MD
H31954.03
ALFAH31954.MDEA
0
SGD
ALFAH31954.MD
ALFAH31954.03
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
1-(3-Pyridylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
CAS No.: 864754-21-2
Molecular Formula: C15H20BN3O2
Formula Weight: 285.16
Melting Point: 102-106°
MDL No.: MFCD08706029
Formula:
C₁₅H₂₀BN₃O₂ MW: 285,16 g/mol Melting Pt: 102…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD08706029 CAS Number: 864754-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...