4-Mercapto-4-methyl-2-pentanol 98%

Supplier: Thermo Scientific
Danger

Synonyms: 4-Mercapto-4-methylpentan-2-ol

H32107.MD H32107.03
ALFAH32107.MDEA 0 SGD
ALFAH32107.MD ALFAH32107.03
4-Mercapto-4-methyl-2-pentanol 98%
4-Mercapto-4-methyl-2-pentanol

Purity: 98% CAS No.: 31539-84-1 Molecular Formula: C6H14OS Formula Weight: 134.24 Flash Point: 70°(158°F) Refractive Index: 1.4710 Boiling Point: 40°/0.6mm MDL No.: MFCD10688294

Formula: C₆H₁₄OS
MW: 134,24 g/mol
Boiling Pt: 40 °C (0,6 mmHg)
Density: 0,957
Flash Pt: 70 °C (158 °F)
Storage Temperature: Ambient
MDL Number: MFCD10688294
CAS Number: 31539-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR