2-Nitro-4-(trifluoromethyl)benzyl chloride 98%
Supplier: Thermo Scientific
|
Danger
|
H32191.03
H32191.09
ALFAH32191.03EA
0
SGD
ALFAH32191.03
ALFAH32191.09
2-Nitro-4-(trifluoromethyl)benzyl chloride 98%
2-Nitro-4-(trifluoromethyl)benzyl chloride
Purity: 98%
CAS No.: 225656-59-7
Molecular Formula: C8H5ClF3NO2
Formula Weight: 239.58
Melting Point: 42-45°
UN No.: UN3261
MDL No.: MFCD01631694
Formula:
C₈H₅ClF₃NO₂ Melting Pt: 42…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD01631694 CAS Number: 225656-59-7 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...