(4-Methoxy-3,5-dimethylphenyl)amine 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Methoxy-3,5-dimethylaniline, 3,5-Dimethyl-4-methoxyaniline

H32232.MD H32232.03
ALFAH32232.MDEA 0 SGD
ALFAH32232.MD ALFAH32232.03
(4-Methoxy-3,5-dimethylphenyl)amine 98%
(4-Methoxy-3,5-dimethylphenyl)amine

Purity: 98% CAS No.: 39785-37-0 Molecular Formula: C9H13NO Formula Weight: 151.21 Melting Point: 62-65° UN No.: UN2811 MDL No.: MFCD01907075

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Melting Pt: 62…65 °C
Storage Temperature: Ambient
MDL Number: MFCD01907075
CAS Number: 39785-37-0
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR